Bis[3-(triethoxysilyl)propyl]tetrasulfide

Supplier: BeanTown Chemical

144930-100G 144930-25G 144930-5G
BT144930-100GEA 271 USD
BT144930-100G BT144930-25G BT144930-5G
Bis[3-(triethoxysilyl)propyl]tetrasulfide
Bis[3-(triethoxysilyl)propyl]tetrasulfide

CAS: 40372-72-3; EC No: 254-896-5; MDL No: MFCD00053751 Liquid; Molecular Formula: C18H42O6S4Si2; MW: 538.95 Flash point: 100°C (212°F) Density (g/mL): 1.08; Refractive Index: 1.49


  • Refractive index: 1.49
  • Liquid

Formula: C₁₈H₄₂O₆S₄Si₂
MW: 538.938 g/mol
Density: 1.08 g/mL
Flash Pt: 100 °C (212 °F)
MDL Number: MFCD00053751
CAS Number: 40372-72-3
EINECS: 254-896-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR