Sodium thiosulfate, anhydrous ≥99%
Supplier: BeanTown Chemical
144885-2.5KG
144885-500G
BT144885-2.5KGEA
126.24
USD
BT144885-2.5KG
BT144885-500G
Sodium thiosulfate, anhydrous ≥99%
Sodium thiosulphate
CAS: 7772-98-7; EC No: 231-867-5; MDL No: MFCD00003499; RTECS: XN6476000
Powder; Linear Formula: Na2S2O3; MW: 158.11
Melting Point: 52° (decomposes)
Density (g/mL): 1.667
Hygroscopic
- RTECS: XN6476000
- Hygroscopic
- Powder
Formula:
Na₂S₂O₃ Melting Pt: 52 °C (dec.) Density: 1.667 g/mL |
MDL Number:
MFCD00003499 CAS Number: 7772-98-7 EINECS: 231-867-5 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...