Thiamine hydrochloride, (max. 5% H₂O) ≥99% (dry weight)

Supplier: BeanTown Chemical

Synonyms: Aneurine hydrochloride, 3-((4-Amino-2-methylpyrimidin-5-yl)methyl)-5-(2-hydroxyethyl)-4-methylthiazol-3-ium chloride hydrochloride, Thiaminium chloride, VitamineB1 hydrochloride

144840-1KG 144840-250G 144840-50G
BT144840-1KGEA 328 USD
BT144840-1KG BT144840-250G BT144840-50G
Thiamine hydrochloride, (max. 5% H₂O) ≥99% (dry weight)
Thiamine hydrochloride

CAS: 67-03-8; EC No: 200-641-8; MDL No: MFCD00012780; RTECS: XI7350000 Crystalline/Powder; Molecular Formula: C12H17ClN4OS·HCl; MW: 337.28 Melting Point: 250° (decomposes) Hygroscopic , Light Sensitive


  • RTECS: XI7350000
  • Hygroscopic, Light Sensitive
  • Crystalline/Powder

Formula: C₁₂H₁₈Cl₂N₄OS
MW: 337.27 g/mol
Melting Pt: 250 °C (dec.)
MDL Number: MFCD00012780
CAS Number: 67-03-8
EINECS: 200-641-8

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR