2-Iodo-3-pyridinol ≥98%

Supplier: BeanTown Chemical

Synonyms: 3-Hydroxy-2-iodopyridine

144775-1G 144775-25G 144775-5G
BT144775-1GEA 65 USD
BT144775-1G BT144775-25G BT144775-5G
2-Iodo-3-pyridinol ≥98%
2-Iodo-3-pyridinol

CAS: 40263-57-8; EC No: 254-864-0; MDL No: MFCD00023421 Solid; Molecular Formula: C5H4INO; MW: 221.00 Melting Point: 196-200° (decomposes) Light Sensitive


  • Light Sensitive
  • Solid

Formula: C₅H₄INO
MW: 221.00 g/mol
Melting Pt: 196…200 °C (dec.)
MDL Number: MFCD00023421
CAS Number: 40263-57-8
EINECS: 254-864-0

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR