Diphenyl carbonate ≥99%
Supplier: BeanTown Chemical
CAS: 102-09-0; EC No: 203-005-8; MDL No: MFCD00003037; RTECS: FG0500000
Flakes; Linear Formula: (C6H5O)2CO; Molecular Formula: C13H10O3; MW: 214.22
Melting Point: 79-82°; Boiling Point: 301-302°; Flash point: 168°C (334°F)
- RTECS: FG0500000
- Flakes
Formula:
C₁₃H₁₀O₃ Boiling Pt: 301…302 °C Melting Pt: 79…82 °C Flash Pt: 168 °C (334 °F) |
MDL Number:
MFCD00003037 CAS Number: 102-09-0 EINECS: 203-005-8 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...