Cystamine dihydrochloride ≥97%
Supplier: BeanTown Chemical
Synonyms:
Cystaminium dichloride, 2,2′-Diaminodiethyl disulphide dihydrochloride, 2,2'-Disulphanediyldiethanamine dihydrochloride, Bis(2-aminoethyl) disulphide dihydrochloride, Decarboxycystine dihydrochloride, Cystamine (dihydrochloride), {2-[(2-Aminoethyl)disulphanyl]ethyl}amine dihydrochloride, 2,2′-Dithiobis(ethylamine) dihydrochloride
144760-100G
144760-25G
BT144760-100GEA
103
USD
BT144760-100G
BT144760-25G
Cystamine dihydrochloride ≥97%
Cystamine dihydrochloride
CAS: 56-17-7; EC No: 200-260-7; MDL No: MFCD00012905; RTECS: KR7260000
Powder; Molecular Formula: C4H12N2S2·2HCl; MW: 225.20
Melting Point: 217-220°
Hygroscopic
- RTECS: KR7260000
- Hygroscopic
- Powder
Formula:
C₄H₁₂N₂S₂·2HCl MW: 225.20 g/mol Melting Pt: 217…220 °C |
MDL Number:
MFCD00012905 CAS Number: 56-17-7 EINECS: 200-260-7 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...