Morpholine-4-carboxamidine hydrobromide ≥98%
Supplier: BeanTown Chemical
Synonyms:
Morpholine-4-carboximidamide hydrobromide
144540-100G
144540-25G
144540-5G
BT144540-100GEA
994
USD
BT144540-100G
BT144540-25G
BT144540-5G
Morpholine-4-carboxamidine hydrobromide ≥98%
Morpholine-4-carboxamidine hydrobromide
CAS: 157415-17-3; MDL No: MFCD00051994
Solid; Molecular Formula: C5H11N3O·HBr; MW: 210.08
Melting Point: 155-165°
Hygroscopic
- Hygroscopic
- Solid
Formula:
C₅H₁₂BrN₃O MW: 210.075 g/mol Melting Pt: 155…165 °C |
MDL Number:
MFCD00051994 CAS Number: 157415-17-3 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...