Humic acid sodium salt, Technical Grade

Supplier: BeanTown Chemical

144535-100G 144535-500G
BT144535-100GEA 46.6 USD
BT144535-100G BT144535-500G
Humic acid sodium salt, Technical Grade
Humic acid sodium salt

CAS: 68131-04-4; EC No: 268-608-0; MDL No: MFCD00135560; RTECS: MT6550000 Powder Melting Point: 300°


  • RTECS: MT6550000
  • Powder

Formula: C₉H₈Na₂O₄
Melting Pt: >300 °C
MDL Number: MFCD00135560
CAS Number: 68131-04-4
EINECS: 268-608-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR