1-Bromo-2-ethylbenzene ≥98%
Supplier: BeanTown Chemical
144510-100G
144510-25G
144510-5G
BT144510-100GEA
419
USD
BT144510-100G
BT144510-25G
BT144510-5G
1-Bromo-2-ethylbenzene ≥98%
1-Bromo-2-ethylbenzene
CAS: 1973-22-4; EC No: 217-823-8; MDL No: MFCD00000077
Liquid; Molecular Formula: C8H9Br; MW: 185.06
Boiling Point: 199°; Flash point: 77°C (171°F)
Density (g/mL): 1.338; Refractive Index: 1.549
- Refractive index: 1.549
- Liquid
Formula:
C₈H₉Br MW: 185.06 g/mol Boiling Pt: 199 °C Density: 1.338 g/mL Flash Pt: 77 °C (171 °F) |
MDL Number:
MFCD00000077 CAS Number: 1973-22-4 EINECS: 217-823-8 UN: 3082 ADR: 9,III |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...