Tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')hafnium(IV) ≥99.5%

Supplier: BeanTown Chemical

Synonyms: Tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato)hafnium(IV)

144500-1G 144500-5G
BT144500-1GEA 107 USD
BT144500-1G BT144500-5G
Tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')hafnium(IV) ≥99.5%
Tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')hafnium(IV)

CAS: 63370-90-1; MDL No: MFCD01863471 Powder; Molecular Formula: C44H80HfO8 ; MW: 915.60 Melting Point: 315°


  • Powder

Formula: C₄₄H₇₆HfO₈
MW: 910.5 g/mol
Melting Pt: 315 °C
MDL Number: MFCD01863471
CAS Number: 63370-90-1

Order Now

Specification Test Results

Assay 99.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR