(R)-(+)-1-Boc-3-methylpiperazine ≥97%
Supplier: BeanTown Chemical
144465-1G
144465-5G
BT144465-1GEA
73
USD
BT144465-1G
BT144465-5G
(R)-(+)-1-Boc-3-methylpiperazine ≥97%
(R)-(+)-1-Boc-3-methylpiperazine
CAS: 163765-44-4; MDL No: MFCD02683205
Crystalline/Solid; Molecular Formula: C10H20N2O2; MW: 200.28
Optical Rotation: [α]/D +16±2°, c = 1 in dioxane
- Optical rotation: [α]/D +16±2°, c = 1 in dioxane
- Crystalline/Solid
Formula:
C₁₀H₂₀N₂O₂ |
MDL Number:
MFCD02683205 CAS Number: 163765-44-4 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...