(±)-α-Terpineol ≥96%

Supplier: BeanTown Chemical

Synonyms: α-Terpineol, Terpineol, 2-(4-Methylcyclohex-3-en-1-yl)propan-2-ol

144440-100G 144440-25G 144440-500G
BT144440-100GEA 40 USD
BT144440-100G BT144440-25G BT144440-500G
(±)-α-Terpineol ≥96%
(±)-α-Terpineol

CAS: 98-55-5; EC No: 202-680-6; MDL No: MFCD00001557; RTECS: WZ6700000 Liquid ; Molecular Formula: C10H18O; MW: 154.24 Melting Point: 31-35°; Boiling Point: 217-218°; Flash point: 90°C (194°F) Density (g/mL): 0.93; Refractive Index: 1.482


  • RTECS: WZ6700000
  • Refractive index: 1.482
  • Liquid

Formula: C₁₀H₁₈O
MW: 154.25 g/mol
Boiling Pt: 217…218 °C
Melting Pt: 31…35 °C
Density: 0.93 g/mL
Flash Pt: 90 °C (194 °F)
MDL Number: MFCD00001557
CAS Number: 98-55-5
EINECS: 202-680-6

Order Now

Specification Test Results

Assay 96%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR