Lithium bromide hydrate ≥99.995%
Supplier: BeanTown Chemical
Synonyms:
Lithium bromide xhydrate
144330-100G
144330-25G
BT144330-100GEA
463.21
USD
BT144330-100G
BT144330-25G
Lithium bromide hydrate ≥99.995%
Lithium bromide hydrate
CAS: 85017-82-9; EC No: 231-439-8; MDL No: MFCD00149763
Crystalline Aggregates; Linear Formula: LiBr·xH2O; MW: 86.85 (anhydrous)
Melting Point: 162-167°
Hygroscopic
- Hygroscopic
- Crystalline Aggregates
Formula:
BrLi·xH₂O MW: 86.85 g/mol (anhy) Boiling Pt: 1265 Melting Pt: 549 |
MDL Number:
MFCD00149763 CAS Number: 85017-82-9 EINECS: 231-439-8 |
Specification Test Results
Assay | 99.995% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...