2-Chloro-4-iodopyridine-3-carboxaldehyde ≥98%

Supplier: BeanTown Chemical

Synonyms: 2-Chloro-4-iodonicotinaldehyde

144325-1G 144325-5G
BT144325-1GEA 205 USD
BT144325-1G BT144325-5G
2-Chloro-4-iodopyridine-3-carboxaldehyde ≥98%
2-Chloro-4-iodopyridine-3-carboxaldehyde

CAS: 153034-90-3; MDL No: MFCD03095294 Solid; Molecular Formula: C6H3ClINO; MW: 267.45 Air Sensitive, Light Sensitive


  • Air Sensitive, Light Sensitive
  • Solid

Formula: C₆H₃ClINO
MW: 267.45 g/mol
MDL Number: MFCD03095294
CAS Number: 153034-90-3

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR