Cesium chloride ≥99.999%

Supplier: BeanTown Chemical

Synonyms: Cesium Chloride

144315-100G 144315-25G
BT144315-100GEA 839.21 USD
BT144315-100G BT144315-25G
Cesium chloride ≥99.999%
Cesium chloride

CAS: 7647-17-8; EC No: 231-600-2; MDL No: MFCD00010955; RTECS: FK9625000 Crystalline/Powder; Linear Formula: CsCl; MW: 168.36 Melting Point: 645° Hygroscopic


  • RTECS: FK9625000
  • Hygroscopic
  • Crystalline/Powder

Formula: CsCl
MW: 168.36 g/mol
Boiling Pt: 1303
Melting Pt: 646
MDL Number: MFCD00010955
CAS Number: 7647-17-8
EINECS: 231-600-2
Merck Index: 13,02021

Order Now

Specification Test Results

Assay 99.999%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR