Boc-L-tyrosine methyl ester ≥99%
Supplier: BeanTown Chemical
144295-100G
144295-25G
144295-5G
BT144295-100GEA
936
USD
BT144295-100G
BT144295-25G
BT144295-5G
Boc-L-tyrosine methyl ester ≥99%
Boc-L-tyrosine methyl ester
CAS: 4326-36-7; MDL No: MFCD00191181
Solid; Molecular Formula: C15H21NO5 ; MW: 295.33
Melting Point: 100-104°
Optical Rotation: [α]22/D +51°, c = 1 in chloroform
- Optical rotation: [α]22/D +51°, c = 1 in chloroform
- Solid
Formula:
C₁₅H₂₁NO₅ Melting Pt: 100…104 °C |
MDL Number:
MFCD00191181 CAS Number: 4326-36-7 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...