Hafnium dioxide ≥99.9% (trace metals basis excluding Zr)

Supplier: BeanTown Chemical

144280-1G 144280-25G 144280-5G
BT144280-1GEA 29 USD
BT144280-1G BT144280-25G BT144280-5G
Hafnium dioxide ≥99.9% (trace metals basis excluding Zr)
Hafnium dioxide

CAS: 12055-23-1; EC No: 235-013-2; MDL No: MFCD00003565 3-12mm Sintered Lump; Molecular Formula: HfO2; MW: 210.49 Density (g/mL): 9.68


  • 3-12mm Sintered Lump

Formula: HfO₂
MW: 210.49 g/mol
Density: 9.68 g/mL
MDL Number: MFCD00003565
CAS Number: 12055-23-1
EINECS: 235-013-2

Order Now

Specification Test Results

Assay 99.9%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR