D-(+)-Proline ≥99%
Supplier: BeanTown Chemical
Synonyms:
H-D-Pro-OH, (R)-(+)-Proline, D-Proline
144240-1G
144240-25G
144240-5G
BT144240-1GEA
40
USD
BT144240-1G
BT144240-25G
BT144240-5G
D-(+)-Proline ≥99%
D-(+)-Proline
CAS: 344-25-2; EC No: 206-452-7; MDL No: MFCD00064317
Powder; Molecular Formula: C5H9NO2; MW: 115.13
Melting Point: 223° (decomposes)
Optical Rotation: [α]22/D +85.0°, c = 4 in water
Hygroscopic
- Optical rotation: [α]22/D +85.0°, c = 4 in water
- Hygroscopic
- Powder
Formula:
C₅H₉NO₂ MW: 115.13 g/mol Melting Pt: 223 °C (dec.) |
MDL Number:
MFCD00064317 CAS Number: 344-25-2 EINECS: 206-452-7 |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...