3-Methoxyphenylboronic acid ≥97%

Supplier: BeanTown Chemical

Synonyms: (3-Methoxyphenyl)metaboric acid, Dihydroxy(3-methoxyphenyl)borane, 3-Methoxybenzeneboronic acid

144215-1G 144215-25G 144215-5G
BT144215-1GEA 37 USD
BT144215-1G BT144215-25G BT144215-5G
3-Methoxyphenylboronic acid ≥97%
3-Methoxyphenylboronic acid

CAS: 10365-98-7; MDL No: MFCD00161359 Solid; Molecular Formula: C7H9BO3; MW: 151.96 Melting Point: 160-163°


  • Solid

Formula: C₇H₉BO₃
MW: 151.96 g/mol
Melting Pt: 160…163 °C
MDL Number: MFCD00161359
CAS Number: 10365-98-7

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR