2'-Chloro-6'-fluoro-3'-methylacetophenone ≥97%

Supplier: BeanTown Chemical

Synonyms: 1-(2-Chloro-6-fluoro-3-methylphenyl)ethanone

144170-1G 144170-5G
BT144170-1GEA 83 USD
BT144170-1G BT144170-5G
2'-Chloro-6'-fluoro-3'-methylacetophenone ≥97%
2'-Chloro-6'-fluoro-3'-methylacetophenone

CAS: 261762-63-4; MDL No: MFCD01631368 Liquid; Molecular Formula: C9H8ClFO; MW: 186.61


  • Liquid

Formula: C₉H₈ClFO
MW: 186.61 g/mol
MDL Number: MFCD01631368
CAS Number: 261762-63-4

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR