Magnesium silicate ≥99% (trace metals basis)

Supplier: BeanTown Chemical

Synonyms: Florisil

144135-250G 144135-50G
BT144135-250GEA 858 USD
BT144135-250G BT144135-50G
Magnesium silicate ≥99% (trace metals basis)
Magnesium silicate

CAS: 1343-88-0; EC No: 215-681-1; MDL No: MFCD00078249 -350 Mesh Powder; Molecular Formula: Mg2O4Si ; MW: 140.71 Melting Point: 1910° Density (g/mL): 0.127


  • -350 Mesh Powder

Formula: MgO₃Si
MW: 100.38 g/mol
Melting Pt: 1910 °C
Density: 0.127 g/mL
MDL Number: MFCD00078249
CAS Number: 1343-88-0
EINECS: 215-681-1

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR