N-(2-Bromoethyl)phthalimide ≥98%
Supplier: BeanTown Chemical
Synonyms:
2-(2-Bromoethyl)isoindoline-1,3-dione
144080-100G
144080-25G
144080-500G
BT144080-100GEA
145
USD
BT144080-100G
BT144080-25G
BT144080-500G
N-(2-Bromoethyl)phthalimide ≥98%
N-(2-Bromoethyl)phthalimide
CAS: 574-98-1; EC No: 209-379-9; MDL No: MFCD00005902
Powder; Molecular Formula: C10H8BrNO2; MW: 254.09
Melting Point: 80-83°
- Powder
Formula:
C₁₀H₈BrNO₂ MW: 254.08 g/mol Melting Pt: 80…83 °C |
MDL Number:
MFCD00005902 CAS Number: 574-98-1 EINECS: 209-379-9 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...