3-Cyano-6-isopropylchromone

Supplier: BeanTown Chemical

144065-1G 144065-5G
BT144065-1GEA 148 USD
BT144065-1G BT144065-5G
3-Cyano-6-isopropylchromone
3-Cyano-6-isopropylchromone

CAS: 50743-32-3; MDL No: MFCD00191961 Solid; Molecular Formula: C13H11NO2; MW: 213.23


  • Solid

Formula: C₁₃H₁₁NO₂
MDL Number: MFCD00191961
CAS Number: 50743-32-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR