Trilithium orthophosphate ≥99.99%

Supplier: BeanTown Chemical

Synonyms: Lithium phosphate tribasic, Lithium orthophosphate, Lithium phosphate

144040-100G 144040-25G
BT144040-100GEA 438 USD
BT144040-100G BT144040-25G
Trilithium orthophosphate ≥99.99%
Trilithium orthophosphate

CAS: 10377-52-3; EC No: 233-823-0; MDL No: MFCD00016187 Powder; Linear Formula: Li3PO4; MW: 115.79


  • Powder

Formula: Li₃O₄P
MW: 115.79
Melting Pt: 1225
Density: 2.41
MDL Number: MFCD00016187
CAS Number: 10377-52-3
EINECS: 233-823-0

Order Now

Specification Test Results

Assay 99.99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR