2-(2-Chloroethoxy)ethanol ≥99%

Supplier: BeanTown Chemical

144020-1KG 144020-250G 144020-50G
BT144020-1KGEA 480 USD
BT144020-1KG BT144020-250G BT144020-50G
2-(2-Chloroethoxy)ethanol ≥99%
2-(2-Chloroethoxy)ethanol

CAS: 628-89-7; EC No: 211-059-9; MDL No: MFCD00002870; RTECS: KK1350000 Liquid; Linear Formula: ClCH2CH2OCH2CH2OH; Molecular Formula: C4H9ClO2 ; MW: 124.57 Boiling Point: 79-81°/5 mmHg; Flash point: 90°C (194°F) Density (g/mL): 1.18; Refractive Index: 1.452


  • RTECS: KK1350000
  • Refractive index: 1.452
  • Liquid

Formula: C₄H₉ClO₂
MW: 124.57 g/mol
Boiling Pt: 79…81 °C/5 mmHg
Density: 1.18 g/mL
Flash Pt: 90 °C (194 °F)
MDL Number: MFCD00002870
CAS Number: 628-89-7
EINECS: 211-059-9
UN: 3334
ADR: 9,

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR