4-Chloro-3-nitropyridine ≥98%
Supplier: BeanTown Chemical
144000-25G
144000-5G
BT144000-25GEA
378
USD
BT144000-25G
BT144000-5G
4-Chloro-3-nitropyridine ≥98%
4-Chloro-3-nitropyridine
CAS: 13091-23-1; MDL No: MFCD02683025
Solid; Molecular Formula: C5H3ClN2O2 ; MW: 158.54
Melting Point: 30-35°
- Solid
Formula:
C₅H₃ClN₂O₂ MW: 158.54 g/mol Melting Pt: 30…35 °C |
MDL Number:
MFCD02683025 CAS Number: 13091-23-1 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...