2-(Methoxymethyl)pyridine

Supplier: BeanTown Chemical

143970-1G 143970-5G
BT143970-1GEA 86 USD
BT143970-1G BT143970-5G
2-(Methoxymethyl)pyridine
2-(Methoxymethyl)pyridine

CAS: 23579-92-2; MDL No: MFCD00030825 Liquid; Molecular Formula: C7H9NO; MW: 123.16


  • Liquid

Formula: C₇H₉NO
MW: 123.1500 g/mol
MDL Number: MFCD00030825
CAS Number: 23579-92-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR