2-(4-Chlorophenyl)ethanol ≥99%

Supplier: BeanTown Chemical

Synonyms: 4-Chlorophenethyl alcohol

143955-10G 143955-50G
BT143955-10GEA 114 USD
BT143955-10G BT143955-50G
2-(4-Chlorophenyl)ethanol ≥99%
2-(4-Chlorophenyl)ethanol

CAS: 1875-88-3; EC No: 217-506-4; MDL No: MFCD00002899 Liquid; Linear Formula: C6H4(Cl)CH2CH2OH; Molecular Formula: C8H9ClO; MW: 156.61 Boiling Point: 110°/0.5 mmHg; Flash point: 110°C (230°F) Density (g/mL): 1.157; Refractive Index: 1.548


  • Refractive index: 1.548
  • Liquid

Formula: C₈H₉ClO
MW: 156.61 g/mol
Boiling Pt: 110 °C/0.5 mmHg
Density: 1.157 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00002899
CAS Number: 1875-88-3
EINECS: 217-506-4

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR