3-Acetyl-2,5-dichlorothiophene ≥98%

Supplier: BeanTown Chemical

Synonyms: 1-(2,5-Dichloro-3-thienyl)-1-ethanone

143920-100G 143920-25G 143920-5G
BT143920-100GEA 978 USD
BT143920-100G BT143920-25G BT143920-5G
3-Acetyl-2,5-dichlorothiophene ≥98%
3-Acetyl-2,5-dichlorothiophene

CAS: 36157-40-1; EC No: 252-893-3; MDL No: MFCD00014522 Crystalline/Powder; Molecular Formula: C6H4Cl2OS; MW: 195.07 Melting Point: 37-40°; Flash point: 110°C (230°F)


  • Crystalline/Powder

Formula: C₆H₄Cl₂OS
MW: 195.07 g/mol
Melting Pt: 37…40 °C
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00014522
CAS Number: 36157-40-1
EINECS: 252-893-3

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR