D-(+)-Tyrosine ≥99%
Supplier: BeanTown Chemical
Synonyms:
H-D-Tyr-OH
143865-1G
143865-25G
143865-5G
BT143865-1GEA
20.99
USD
BT143865-1G
BT143865-25G
BT143865-5G
D-(+)-Tyrosine ≥99%
D-(+)-Tyrosine
CAS: 556-02-5; EC No: 209-112-6; MDL No: MFCD00063073
Powder; Molecular Formula: C9H11NO3; MW: 181.19
Optical Rotation: [α]20/D +10.3°, c = 5 in 1M HCl
- Optical rotation: [α]20/D +10.3°, c = 5 in 1M HCl
- Powder
Formula:
C₉H₁₁NO₃ MW: 181.19 g/mol |
MDL Number:
MFCD00063073 CAS Number: 556-02-5 EINECS: 209-112-6 |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...