1-Adamantanecarboxamide ≥97%
Supplier: BeanTown Chemical
143860-1G
143860-5G
BT143860-1GEA
58
USD
BT143860-1G
BT143860-5G
1-Adamantanecarboxamide ≥97%
1-Adamantanecarboxamide
CAS: 5511-18-2; EC No: 234-074-2; MDL No: MFCD00077200; RTECS: AU4452000
Solid; Molecular Formula: C11H17NO; MW: 179.26
Melting Point: 188-190°
- RTECS: AU4452000
- Solid
Formula:
C₁₁H₁₇NO Melting Pt: 188…190 °C |
MDL Number:
MFCD00077200 CAS Number: 5511-18-2 EINECS: 234-074-2 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...