N-Boc-L-alanine ≥98%
Supplier: BeanTown Chemical
Synonyms:
N-Alpha-t-Boc-L-alanine, (S)-2-((tert-Butoxycarbonyl)amino)propanoic acid, BOC-L-Alanine, Boc-Ala-OH
143845-25G
143845-5G
BT143845-25GEA
100
USD
BT143845-25G
BT143845-5G
N-Boc-L-alanine ≥98%
N-Boc-L-alanine
CAS: 15761-38-3; EC No: 239-847-8; MDL No: MFCD00037225
Powder; Linear Formula: CH3CH[NHCO2C(CH3)3]COOH; Molecular Formula: C8H15NO4; MW: 189.21
Melting Point: 79-83°
Optical Rotation: [α]20/D -25±1°, c = 2 in acetic acid
- Optical rotation: [α]20/D -25±1°, c = 2 in acetic acid
- Powder
Formula:
C₈H₁₅NO₄ MW: 189.21 g/mol Melting Pt: 79…83 °C |
MDL Number:
MFCD00037225 CAS Number: 15761-38-3 EINECS: 239-847-8 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...