(2-Carboxyethyl)phenylphosphinic acid
Supplier: BeanTown Chemical
Synonyms:
3-(Hydroxy(phenyl)phosphoryl)propanoic acid, 3-Hydroxyphenylphosphinylpropanoic acid
143825-1G
143825-5G
BT143825-1GEA
74
USD
BT143825-1G
BT143825-5G
(2-Carboxyethyl)phenylphosphinic acid
(2-Carboxyethyl)phenylphosphinic acid
CAS: 14657-64-8; EC No: 411-200-6; MDL No: MFCD08704562
Solid; Molecular Formula: C9H11O4P; MW: 214.16
- Solid
Formula:
C₉H₁₁O₄P MW: 214.16 g/mol |
MDL Number:
MFCD08704562 CAS Number: 14657-64-8 EINECS: 411-200-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...