Tungsten, gauze woven from wire 30 mesh Ø 0.1018 mm
Supplier: BeanTown Chemical
143795-150X150
143795-300X300
143795-75X75MM
BT143795-150X1EA
334
USD
BT143795-150X1
BT143795-300X3
BT143795-75X75
Tungsten, gauze woven from wire 30 mesh Ø 0.1018 mm
Tungsten
CAS: 7440-33-7; EC No: 231-143-9; MDL No: MFCD00011461
Wire Cloth; Molecular Formula: W; MW: 183.84
Melting Point: 3410°; Boiling Point: 5660°
Density (g/mL): 19.3
- Wire Cloth
Formula:
W MW: 183.84 g/mol Boiling Pt: 5660 Melting Pt: 3410 Density: 19.3 g/mL |
MDL Number:
MFCD00011461 CAS Number: 7440-33-7 EINECS: 231-143-9 Merck Index: 12,09945 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...