α,α,α-Trifluoro-4-tolualdehyde ≥97%

Supplier: BeanTown Chemical

Synonyms: 4-(Trifluoromethyl)benzaldehyde, 1-Formyl-4-trifluoromethylbenzene, p-Trifluoromethylbenzaldehyde

143785-25G 143785-5G
BT143785-25GEA 146 USD
BT143785-25G BT143785-5G
α,α,α-Trifluoro-4-tolualdehyde ≥97%
α,α,α-Trifluoro-4-tolualdehyde

CAS: 455-19-6; EC No: 207-240-7; MDL No: MFCD00006952 Liquid; Molecular Formula: C8H5F3O; MW: 174.12 Boiling Point: 66-67°/13 mmHg; Flash point: 65°C (149°F) Density (g/mL): 1.275; Refractive Index: 1.463 Air Sensitive


  • Refractive index: 1.463
  • Air Sensitive
  • Liquid

Formula: C₈H₅F₃O
MW: 174.12 g/mol
MDL Number: MFCD00006952
CAS Number: 455-19-6

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR