Bis(triphenylphosphine)iminium trifluoroacetate ≥98%

Supplier: BeanTown Chemical

143770-25G 143770-5G
BT143770-25GEA 352 USD
BT143770-25G BT143770-5G
Bis(triphenylphosphine)iminium trifluoroacetate ≥98%
Bis(triphenylphosphine)iminium trifluoroacetate

CAS: 116405-43-7 Solid; Molecular Formula: C36H30NP2·C2F3O2; MW: 782.65


  • Solid

Formula: C₃₈H₃₀F₃NO₂P₂
MDL Number: MFCD15146355
CAS Number: 116405-43-7

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR