1,1-Dimethylurea ≥98%
Supplier: BeanTown Chemical
Synonyms:
N,N-Dimethylurea
143755-10G
143755-50G
BT143755-10GEA
44
USD
BT143755-10G
BT143755-50G
1,1-Dimethylurea ≥98%
1,1-Dimethylurea
CAS: 598-94-7; EC No: 202-498-7; MDL No: MFCD00007959; RTECS: YS9867985
Crystalline; Linear Formula: (CH3)2NCONH2; Molecular Formula: C3H8N2O; MW: 88.11
Melting Point: 178-183°
- RTECS: YS9867985
- Crystalline
Formula:
C₃H₈N₂O MW: 88.11 g/mol Melting Pt: 178…183 °C |
MDL Number:
MFCD00007959 CAS Number: 598-94-7 EINECS: 202-498-7 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...