Polyethylene glycol 12,000

Supplier: BeanTown Chemical

Synonyms: PEG 12,000

143735-1KG 143735-250G
BT143735-1KGEA 113 USD
BT143735-1KG BT143735-250G
Polyethylene glycol 12,000
Polyethylene glycol 12,000

CAS: 25322-68-3; MDL No: MFCD00081839 Powder; Linear Formula: H(OCH2CH2)nOH; MW ≈12,000 Melting Point: 64-65°; Flash point: 229°C (444°F)


  • Powder

Formula: H(OCH₂CH₂)nOH
Melting Pt: 64…65 °C
Flash Pt: 229 °C (444 °F)
MDL Number: MFCD00081839
CAS Number: 25322-68-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR