Strontium titanate ≥99%
Supplier: BeanTown Chemical
143710-1KG
143710-250G
BT143710-1KGEA
106
USD
BT143710-1KG
BT143710-250G
Strontium titanate ≥99%
Strontium titanate
CAS: 12060-59-2; EC No: 235-044-1; MDL No: MFCD00049554
Powder; Linear Formula: SrTiO3; MW: 183.50
Melting Point: 2060°
Density (g/mL): 4.810
- Powder
Formula:
O₃SrTi MW: 183.5 Melting Pt: 2080 Density: 4.810 g/mL |
MDL Number:
MFCD00049554 CAS Number: 12060-59-2 EINECS: 235-044-1 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...