1-Adamantanemethanol ≥98%
Supplier: BeanTown Chemical
Synonyms:
1-(Hydroxymethyl)adamantane, 1-(Hydroxymethyl)tricyclo[3.3.1.1(3,7)]decane, Adamantan-1-ylmethanol, 1-Adamantylmethanol
143700-1G
143700-25G
143700-5G
BT143700-1GEA
17
USD
BT143700-1G
BT143700-25G
BT143700-5G
1-Adamantanemethanol ≥98%
1-Adamantanemethanol
CAS: 770-71-8; EC No: 212-225-3; MDL No: MFCD00074751
Crystalline; Molecular Formula: C11H18O; MW: 166.26
Melting Point: 114-117°
- Crystalline
Formula:
C₁₁H₁₈O MW: 166.26 g/mol Melting Pt: 114…117 °C |
MDL Number:
MFCD00074751 CAS Number: 770-71-8 EINECS: 212-225-3 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...