Hafnium(IV) acetylacetonate ≥97%
Supplier: BeanTown Chemical
Synonyms:
Hafnium(IV) 2,4-pentanedionate
143675-1G
143675-25G
143675-5G
BT143675-1GEA
40
USD
BT143675-1G
BT143675-25G
BT143675-5G
Hafnium(IV) acetylacetonate ≥97%
Hafnium (IV) acetylacetonate
CAS: 17475-67-1; MDL No: MFCD00044997
Powder; Molecular Formula: C20H28O8Hf; MW: 574.93
Melting Point: 193°
- Powder
Formula:
C₂₀H₂₈O₈Hf Melting Pt: 193 °C |
MDL Number:
MFCD00044997 CAS Number: 17475-67-1 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...