3-Formyl-2-methoxypyridine ≥97%

Supplier: BeanTown Chemical

143660-1G 143660-5G
BT143660-1GEA 159 USD
BT143660-1G BT143660-5G
3-Formyl-2-methoxypyridine ≥97%
3-Formyl-2-methoxypyridine

CAS: 71255-09-9; MDL No: MFCD04115112 Liquid; Molecular Formula: C7H7NO2; MW: 137.14 Boiling Point: 200-201°; Flash point: 96°C (205°F) Density (g/mL): 1.161; Refractive Index: 1.550 Air Sensitive


  • Refractive index: 1.550
  • Air Sensitive
  • Liquid

Formula: C₇H₇NO₂
Boiling Pt: 200…201 °C
Density: 1.161 g/mL
Flash Pt: 96 °C (205 °F)
MDL Number: MFCD04115112
CAS Number: 71255-09-9

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR