(R)-(-)-Methyl mandelate ≥99%
Supplier: BeanTown Chemical
Synonyms:
D(-)-Methyl glycolate, (R)-(-)-Methyl glycolate, (R)-(-)-Mandelic acid methyl ester, D(-)-Methyl mandelate, D(-)-2-Hydroxy-2-phenylacetic acid methyl ester, D(-)-Mandelic acid methyl ester, (R)-Methyl 2-hydroxy-2-phenylacetate, Methyl (R)-(-)-mandelate, (R)-(-)-2-Hydroxy-2-phenylacetic acid methyl ester
143645-1G
143645-5G
BT143645-1GEA
32
USD
BT143645-1G
BT143645-5G
(R)-(-)-Methyl mandelate ≥99%
(R)-(-)-Methyl mandelate
CAS: 20698-91-3; MDL No: MFCD00064247
Powder; Molecular Formula: C9H10O3 ; MW: 166.17
Melting Point: 56-58°
Optical Rotation: [α]20/D -144°, c = 1 in methanol
- Optical rotation: [α]20/D -144°, c = 1 in methanol
- Powder
Formula:
C₉H₁₀O₃ MW: 166.18 g/mol Melting Pt: 56…58 °C |
MDL Number:
MFCD00064247 CAS Number: 20698-91-3 |
Specification Test Results
Assay | 99% |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...