3-Chloroisonicotinic acid

Supplier: BeanTown Chemical

Synonyms: 3-Chloropyridine-4-carboxylic acid

143615-1G 143615-5G
BT143615-1GEA 25 USD
BT143615-1G BT143615-5G
3-Chloroisonicotinic acid
3-Chloroisonicotinic acid

CAS: 88912-27-0; MDL No: MFCD03094685 Solid; Molecular Formula: C6H4ClNO2 ; MW: 157.55 Melting Point: 227-231° (decomposes)


  • Solid

Formula: C₆H₄ClNO₂
MW: 157.56 g/mol
Melting Pt: 227…231 °C (dec.)
MDL Number: MFCD03094685
CAS Number: 88912-27-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR