6-Bromoindole-3-carboxaldehyde ≥97%

Supplier: BeanTown Chemical

143610-1G 143610-5G
BT143610-1GEA 97 USD
BT143610-1G BT143610-5G
6-Bromoindole-3-carboxaldehyde ≥97%
6-Bromoindole-3-carboxaldehyde

CAS: 17826-04-9; MDL No: MFCD00792689 Solid; Molecular Formula: C9H6BrNO; MW: 224.06 Melting Point: 202-206°


  • Solid

Formula: C₉H₆BrNO
Melting Pt: 202…206 °C
MDL Number: MFCD00792689
CAS Number: 17826-04-9

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR