4-Chloro-3-nitrobenzonitrile ≥98%

Supplier: BeanTown Chemical

143575-25G 143575-5G
BT143575-25GEA 169 USD
BT143575-25G BT143575-5G
4-Chloro-3-nitrobenzonitrile ≥98%
4-Chloro-3-nitrobenzonitrile

CAS: 939-80-0; EC No: 213-364-2; MDL No: MFCD00016987; RTECS: DI3050000 Crystalline; Molecular Formula: C7H3ClN2O2; MW: 182.56 Melting Point: 98-100°


  • RTECS: DI3050000
  • Crystalline

Formula: C₇H₃ClN₂O₂
MW: 182.57 g/mol
Melting Pt: 98…100 °C
MDL Number: MFCD00016987
CAS Number: 939-80-0
EINECS: 213-364-2

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR