2,4-Dichlorobenzyl alcohol ≥99%
Supplier: BeanTown Chemical
Synonyms:
(2,4-Dichlorophenyl)methanol
143570-100G
143570-25G
143570-500G
BT143570-100GEA
174
USD
BT143570-100G
BT143570-25G
BT143570-500G
2,4-Dichlorobenzyl alcohol ≥99%
2,4-Dichlorobenzyl alcohol
CAS: 1777-82-8; EC No: 217-210-5; MDL No: MFCD00004606; RTECS: DO0890000
Crystalline/Powder; Molecular Formula: C7H6Cl2O ; MW: 177.03
Melting Point: 55-58°
- RTECS: DO0890000
- Crystalline/Powder
Formula:
C₇H₆Cl₂O MW: 177.03 g/mol Melting Pt: 55…58 °C |
MDL Number:
MFCD00004606 CAS Number: 1777-82-8 EINECS: 217-210-5 |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...