Chloro(1,5-cyclooctadiene)iridium(I) dimer

Supplier: BeanTown Chemical

Synonyms: [Ir(cod)Cl]2, Chloro-1,5-cyclooctadiene iridium(I) dimer

143565-1G 143565-250MG 143565-5G
BT143565-1GEA 360 USD
BT143565-1G BT143565-250MG BT143565-5G
Chloro(1,5-cyclooctadiene)iridium(I) dimer
Chloro(1,5-cyclooctadiene)iridium (I) dimer

CAS: 12112-67-3; EC No: 235-170-7; MDL No: MFCD00012414 Crystalline/Powder; Molecular Formula: C16H24Cl2Ir2; MW: 671.71 Melting Point: 205° (decomposes)


  • Crystalline/Powder

Formula: C₁₆H₂₄Cl₂Ir₂
MW: 671.70 g/mol
Melting Pt: 205 °C (dec.)
MDL Number: MFCD00012414
CAS Number: 12112-67-3
EINECS: 235-170-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR