Pentaerythritol ≥98%

Supplier: BeanTown Chemical

143545-1KG 143545-250G 143545-5KG
BT143545-1KGEA 42.14 USD
BT143545-1KG BT143545-250G BT143545-5KG
Pentaerythritol ≥98%
Pentaerythritol

CAS: 115-77-5; EC No: 204-104-9; MDL No: MFCD00004692; RTECS: RZ2490000 Powder; Linear Formula: C(CH2OH)4; MW: 136.15 Melting Point: 253-258°; Boiling Point: 276°/30 mmHg; Flash point: 150°C (302°F) Density (g/mL): 1.396


  • RTECS: RZ2490000
  • Powder

Formula: C₅H₁₂O₄
MW: 136.15 g/mol
Boiling Pt: 276 °C/30 mmHg
Melting Pt: 253…258 °C
Density: 1.396 g/mL
Flash Pt: >150 °C (>302 °F)
MDL Number: MFCD00004692
CAS Number: 115-77-5
EINECS: 204-104-9

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR