2-Phenylimidazole ≥98%
Supplier: BeanTown Chemical
Synonyms:
2-Phenyl-1H-imidazole
143525-100G
143525-25G
BT143525-100GEA
99
USD
BT143525-100G
BT143525-25G
2-Phenylimidazole ≥98%
2-Phenylimidazole
CAS: 670-96-2; EC No: 211-581-7; MDL No: MFCD00005186
Solid; Molecular Formula: C9H8N2; MW: 144.17
Melting Point: 142-148°
- Solid
Formula:
C₉H₈N₂ MW: 144.18 g/mol Melting Pt: 142…148 °C |
MDL Number:
MFCD00005186 CAS Number: 670-96-2 EINECS: 211-581-7 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...